TES, 99%
TES, 99%
TES, 99%
Thermo Scientific Chemicals
TES, 99%
CAS: 7365-44-8 | C6H15NO6S | 229.247 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21819.1850 g
Catalog number ALFB21819.18
Price (MYR)
641.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
641.00
EA
Add to cart
Specifications
Chemical Name or MaterialTES
CAS7365-44-8
Melting Point∼225°C (decomposition)
Recommended StorageAmbient temperatures
EINECS Number230-906-3
View more
TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Biological buffer useful in pH range 7.0 - 8.0: Biochemistry, 5, 467 (1966).
  2. Gupta, B. S.; Taha, M.; Lee, M. J. Interactions of bovine serum albumin with biological buffers, TES, TAPS, and TAPSO in aqueous solutions. Process Biochem. 2013, 48 (11), 1686-1696.
  3. Daub, M. E.; Prudhomme, J.; Roch, K. L.; Vanderwal, C. D. Synthesis and Potent Antimalarial Activity of Kalihinol B. J. Am. Chem. Soc. 2015, 137 (15), 4912-4915.