1,6-Dichlorohexane, 98%
1,6-Dichlorohexane, 98%
1,6-Dichlorohexane, 98%
1,6-Dichlorohexane, 98%
Thermo Scientific Chemicals
1,6-Dichlorohexane, 98%
CAS: 2163-00-0 | C6H12Cl2 | 155.062 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB21763.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material1,6-Dichlorohexane
CAS2163-00-0
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c