3-Chloropropionic acid, 98%
3-Chloropropionic acid, 98%
3-Chloropropionic acid, 98%
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB21744.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Chloropropionic acid
CAS107-94-8
Health Hazard 1H314
Health Hazard 2GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more