Isobutyramide, 99%
Isobutyramide, 99%
Isobutyramide, 99%
Thermo Scientific Chemicals
Isobutyramide, 99%
CAS: 563-83-7 | C4H9NO | 87.122 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21641.1425 g
Catalog number ALFB21641.14
Price (MYR)
478.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
478.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsobutyramide
CAS563-83-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Melting Point127°C to 129°C
View more
Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Incompatible with acids, bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Massing, J. O.; Planalp, R. P. Influence of PNIPAm on log K f of a copolymerized 2, 2'-bipyridine: revised bifunctional ligand design for ratiometric metal-ion sensing. Dalton Trans. 2015, 44 (26), 11887-11892.
  2. Kozole, J.; Levine, L. A.; Phillips, J. T.; Stairs, J. R. Gas phase ion chemistry of an ion mobility spectrometry based explosive trace detector elucidated by tandem mass spectrometry. Talanta 2015, 140, 10-19.