2-Chlorobutane, 98+%
2-Chlorobutane, 98+%
2-Chlorobutane, 98+%
2-Chlorobutane, 98+%
Thermo Scientific Chemicals
2-Chlorobutane, 98+%
CAS: 78-86-4 | C4H9Cl | 92.566 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number ALFB21586.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or Material2-Chlorobutane
CAS78-86-4
Health Hazard 1H225-H290
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3P210-P233-P234-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P390-P501c
View more
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.
Solubility
Immiscible with water.
Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Saunders, W. H.; Gronert, S. The Competition between Elimination Pathways in the Reactions of a Wide Variety of Bases with 2-Fluoro- and 2-Chlorobutane in the Gas Phase. J. Org. Chem. 2015, 80 (21), 10787-10793.
  2. Benito, J.; Guerrero, H.; Artigas, H.; López, M. C.; Lafuente, C. Refractive indices and static permittivities of systems containing n-hexane or n-heptane and isomeric chlorobutanes. J. Chem. Thermodyn. 2015, 86, 162-167.