Diethyl oxalacetate sodium salt, 95%
Diethyl oxalacetate sodium salt, 95%
Diethyl oxalacetate sodium salt, 95%
Diethyl oxalacetate sodium salt, 95%
Thermo Scientific Chemicals
Diethyl oxalacetate sodium salt, 95%
CAS: 40876-98-0 | C8H11NaO5 | 210.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB21486.36
Price (MYR)
1,233.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,233.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiethyl oxalacetate sodium salt
CAS40876-98-0
Health Hazard 1H302-H311-H319
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
Melting Point∼185°C
View more
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water (1.2 g/L) (25°C).
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. M F Utter, et al. Pyruvate Carboxylase. I. Nature Of The Reaction.J. Biol. Chem.19632382603-2608.
  2. Charles M. Hall, et al. Quinoline derivatives as antiallergy agents.J. Med. Chem., 197417(7), 685-690.