L-(+)-Asparagine, 99%
L-(+)-Asparagine, 99%
L-(+)-Asparagine, 99%
L-(+)-Asparagine, 99%
Thermo Scientific Chemicals
L-(+)-Asparagine, 99%
CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2500 g
Catalog number ALFB21473.22
Price (MYR)
531.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
531.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-(+)-Asparagine
Melting Point234°C to 236°C (decomposition)
CAS70-47-3
Recommended StorageAmbient temperatures
EINECS Number200-735-9
View more
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.
Solubility
Soluble in acids, bases. Insoluble in methanol, ethanol, ether and benzene.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhang, J.; Guo, Y.; Lu, T.; Shi, H.; Dai, T.; Shi, T. Oxidation of L-asparagine by bis(hydrogenperiodato)argentate(III): a kinetic investigation. Transition Met. Chem. 2015, 40 (3), 281-286.
  2. Asghar, B. H.; Altass, H. M.; Fawzy, A. Transition metal-catalyzed oxidation of L-asparagine by platinum(IV) in acid medium: a kinetic and mechanistic study. Trans. Metal Chem. 2015, 40 (6), 587-594.