4-tert-Butylcalix[8]arene, 96%
4-tert-Butylcalix[8]arene, 96%
4-tert-Butylcalix[8]arene, 96%
4-tert-Butylcalix[8]arene, 96%
Thermo Scientific Chemicals
4-tert-Butylcalix[8]arene, 96%
CAS: 68971-82-4 | C88H112O8 | 1297.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB21443.14
Price (MYR)
1,446.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,446.00
EA
Add to cart
Specifications
Chemical Name or Material4-tert-Butylcalix[8]arene
CAS68971-82-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c