Tetracycline hydrochloride, 96%
Tetracycline hydrochloride, 96%
Tetracycline hydrochloride, 96%
Tetracycline hydrochloride, 96%
Thermo Scientific Chemicals
Tetracycline hydrochloride, 96%
Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria. | CAS: 64-75-5 | C22H25ClN2O8 | 480.898 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB21408.22
Price (MYR)
1,140.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,140.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetracycline hydrochloride
TypeTetracycline Hydrochloride
CAS64-75-5
Health Hazard 1H319-H361d
Health Hazard 2GHS H Statement
H361-H319
Suspected of damaging fertility or the unborn child.
Causes serious eye irritation.
View more
Tetracycline hydrochloride is used to induce apoptosis in osteoclasts. It is used to treat acne and other skin infections, respiratory tract infections like pneumonia, genital, urinary infections, leptospirosis, helicobacter pylori, taxoplasmosis, mycoplasma, psittacosis for dog and cats. It also works more effectively in animals that have tick-borne infections. It is also useful in cell culture applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria
Applications
  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
RUO – Research Use Only
General References:
  1. Chopra, I.; Roberts, M. Tetracycline antibiotics: mode of action, applications, molecular biology, and epidemiology of bacterial resistance. Microbiol Mol Biol Rev. 2001, 65(2), 232-60.
  2. Kenny, P.A.; Enver, T., Ashworth, A. Retroviral vectors for establishing tetracycline-regulated gene expression in an otherwise recalcitrant cell line. BMC Mol Biol. 2002, 3:13.