1,4-Dichlorobutane, 97%
1,4-Dichlorobutane, 97%
1,4-Dichlorobutane, 97%
1,4-Dichlorobutane, 97%
Thermo Scientific Chemicals
1,4-Dichlorobutane, 97%
CAS: 110-56-5 | C4H8Cl2 | 127.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21337.30250 g
Catalog number ALFB21337.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Dichlorobutane
CAS110-56-5
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in organic synthesis including adiponitrile. It is also used as an active pharmaceutical intermediate. 1,4-dichlorobutane (1,4-DCB) can be used as an organic solvent (O) for ion-transfer voltammetry at water (W) interface.
Solubility
Soluble in water at 20°C2.4g/L.
Notes
Store away from oxidizing agents, alkali metals and alkali earth metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Hajime Katano, et al. Ion-transfer voltammetry at 1,6-dichlorohexane∣water and 1,4-dichlorobutane∣water interfaces.Talanta,200463(1), 185-193.
  2. Kirsten Aarset, et al. Molecular Structure and Conformational Composition of 1,4-Dichlorobutane and 1,4-Dibromobutane: a Gas-Phase Electron Diffraction and ab Initio Investigation.J. Phys. Chem. 199498(20), 5249-5256.