Cesium formate, 98%
Cesium formate, 98%
Cesium formate, 98%
Thermo Scientific Chemicals
Cesium formate, 98%
CAS: 3495-36-1 | CH2CsO2 | 178.93 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21327.1425 g
Catalog number ALFB21327.14
Price (MYR)
508.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
508.00
EA
Add to cart
Specifications
Chemical Name or MaterialCesium formate
CAS3495-36-1
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362