2-Acetylbenzo[b]thiophene, 98%
2-Acetylbenzo[b]thiophene, 98%
2-Acetylbenzo[b]thiophene, 98%
2-Acetylbenzo[b]thiophene, 98%
Thermo Scientific Chemicals
2-Acetylbenzo[b]thiophene, 98%
CAS: 22720-75-8 | C10H8OS | 176.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFB21173.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Acetylbenzo[b]thiophene
CAS22720-75-8
Melting Point86°C to 88°C
Recommended StorageAmbient temperatures
EINECS Number245-177-7