Giemsa Stain
Giemsa Stain
Giemsa Stain
Thermo Scientific Chemicals
Giemsa Stain
CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFB21172.18
Price (MYR)
772.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
772.00
EA
Add to cart
Specifications
Chemical Name or MaterialGiemsa Stain
CAS51811-82-6
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
Melting Point∼300°C
View more
Giemsa Stain is used as a laboratory reagent and stain for microscopy. It is also used for the histopathologic detection of malaria and other parasites such as spirochete and protozoan. The stain is also been incorporated in differential staining between bacterial and human cells. The compound has been used in combination with May-Grunwald or Wright Stain (sc-203763) as a histological hematology stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Giemsa Stain is used as a laboratory reagent and stain for microscopy. It is also used for the histopathologic detection of malaria and other parasites such as spirochete and protozoan. The stain is also been incorporated in differential staining between bacterial and human cells. The compound has been used in combination with May-Grunwald or Wright Stain (sc-203763) as a histological hematology stain.
Solubility
Insoluble in water (<0.1%), methanol (5.8 mg/L), and ethanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. K. K. Friend.; S. Chen.; F. H. Ruddle. Differential staining of interspecific chromosomes in somatic cell hybrids by alkaline Giemsa stain. Somatic Cell Genetics. 1976, 2 (2), 183-188.
  2. Julie R. Korenberg.; Elizabeth F. Freedlender. Giemsa technique for the detection of sister chromatid exchanges. Ind. Eng. Chem. Res. 1974, 48 (4), 355-360.