Methyl Yellow, indicator
Methyl Yellow, indicator
Methyl Yellow, indicator
Methyl Yellow, indicator
Thermo Scientific Chemicals
Methyl Yellow, indicator
CAS: 60-11-7 | C14H15N3 | 225.30 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21145.1425 g
Catalog number ALFB21145.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl Yellow
Name Noteindicator
CAS60-11-7
Health Hazard 1H301-H350
Health Hazard 2GHS H Statement
H301-H351
Toxic if swallowed.
Suspected of causing cancer.
View more
Methyl Yellow used as a pH indicator. It is an acid-base indicator that changes color from below pH 2.9 red to above pH 4.0 yellow and vice versa. Typical use is in the titration of a weak base with a strong acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl Yellow used as a pH indicator. It is an acid-base indicator that changes color from below pH 2.9 red to above pH 4.0 yellow and vice versa. Typical use is in the titration of a weak base with a strong acid.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Transition interval: pH 2.9 (red) to pH 4.0 (yellow)
RUO – Research Use Only
General References:
  1. Makoto Nagase.; Yasuhiko Osaki.; Takahiko Matsueda. Determination of methyl yellow, sudan I and sudan II in water by high-performance liquid chromatography. Journal of Chromatography A. 1989, 465 (3), 434-437.
  2. Ross R. Weikel.; Jason P. Hallett.; Charles L. Liotta.; Charles A. Eckert. Self-Neutralizing in Situ Acid Catalysis for Single-Pot Synthesis of Iodobenzene and Methyl Yellow in CO2-Expanded Methanol. Ind. Eng. Chem. Res. 2007, 46 (16), 5252-5257.