L-(+)-Ribose, 99%, Thermo Scientific Chemicals
L-(+)-Ribose, 99%, Thermo Scientific Chemicals
L-(+)-Ribose, 99%, Thermo Scientific Chemicals
L-(+)-Ribose, 99%, Thermo Scientific Chemicals
L-(+)-Ribose, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB21117.065 g
Catalog number ALFB21117.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-(+)-Ribose
CAS24259-59-4
Melting Point85°C to 88°C
Recommended StorageAmbient temperatures
EINECS Number246-110-4
View more
L-Ribose is a potential starting material for the synthesis of many L-nucleoside-based pharmaceutical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Ribose is a potential starting material for the synthesis of many L-nucleoside-based pharmaceutical compounds.
Solubility
Soluble in water (100 mg/ml).
Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Kazuya Okano. Synthesis and pharmaceutical application of l-ribose.Tetrahedron.200965(10), 1937-1949.
  2. Soo-Jin Yeom, et al. L-Ribose Production from L-Arabinose by Using Purified L-Arabinose Isomerase and Mannose-6-Phosphate Isomerase from Geobacillus thermodenitrificans.Appl. Environ. Microbiol.200975(21), 6941-6943.