N-Fmoc-L-leucine, 98%
N-Fmoc-L-leucine, 98%
N-Fmoc-L-leucine, 98%
N-Fmoc-L-leucine, 98%
Thermo Scientific Chemicals
N-Fmoc-L-leucine, 98%
CAS: 35661-60-0 | C21H23NO4 | 353.42 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21040.1425 g
Catalog number ALFB21040.14
Price (MYR)
587.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
587.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Fmoc-L-leucine
CAS35661-60-0
Melting Point152°C to 155°C
Recommended StorageKeep cold
EINECS Number252-662-7