Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer)
Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer)
Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer)
Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer)
Thermo Scientific Chemicals
Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer)
CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20932.22100 g
Catalog number ALFB20932.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMetaphosphoric acid
CAS37267-86-0
Health Hazard 1H314-H335
Density2.2
Health Hazard 2GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
View more
Metaphosphoric acid is used as a dehydrating agent. Its derivative, sodium hexametaphosphate is used as a sequestrant and a food additive. It is also used as a phosphorylating agent, analytical reagent and in dental cements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Metaphosphoric acid is used as a dehydrating agent. Its derivative, sodium hexametaphosphate is used as a sequestrant and a food additive. It is also used as a phosphorylating agent, analytical reagent and in dental cements.
Solubility
Soluble in alcohol.
Notes
Form: Lumps.Hygroscopic. Moisture sensitive. Incompatible with strong bases, nitromethane and metals.
RUO – Research Use Only
General References:
  1. Alkorta, I.; Azofra, L. M.; Elguero, J. Ab initio study in the hydration process of metaphosphoric acid: the importance of the pnictogen interactions. Theor. Chem. Acc. 2015, 134 (3), 1-8.
  2. Feitosa, V. P.; Bazzocchi, M. G.; Putignano, A.; Orsini, G.; Luzi, A. L.; Sinhoreti, M. A. C.; Watson, T. F.; Sauro, S. Dicalcium phosphate(CaHPO4·2H2O) precipitation through ortho- or meta-phosphoric acid-etching: Effects on the durability and nanoleakage/ultra-morphology of resin-dentine interfaces. J. Dent. 2013, 41 (11), 1068-1080.