View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Decanol
CAS2051-31-2
Health Hazard 1H227
Health Hazard 2GHS H Statement H227 Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.