Tetraethylthiuram disulfide, 97%
Tetraethylthiuram disulfide, 97%
Tetraethylthiuram disulfide, 97%
Tetraethylthiuram disulfide, 97%
Thermo Scientific Chemicals
Tetraethylthiuram disulfide, 97%
CAS: 97-77-8 | C10H20N2S4 | 296.524 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Catalog number ALFB20721.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylthiuram disulfide
CAS97-77-8
Health Hazard 1H302-H317-H373
Health Hazard 2GHS H Statement
H373-H302-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
View more
Dopamine beta-hydroxylase inhibitor