Ethylmalonic acid, 97+%
Ethylmalonic acid, 97+%
Ethylmalonic acid, 97+%
Ethylmalonic acid, 97+%
Thermo Scientific Chemicals
Ethylmalonic acid, 97+%
CAS: 601-75-2 | C5H8O4 | 132.115 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20715.065 g
Catalog number ALFB20715.06
Price (MYR)
309.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
309.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylmalonic acid
CAS601-75-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Elliot R. Alexander.; Elizabeth J. Underhill. Studies on the Mechanism of the Mannich Reaction. I. Ethylmalonic Acid, A Methynyl Compound. J. Am. Chem. Soc. 1949, 71 (12),4014-4019 .