Phenyl salicylate, 99%
Phenyl salicylate, 99%
Phenyl salicylate, 99%
Phenyl salicylate, 99%
Thermo Scientific Chemicals
Phenyl salicylate, 99%
CAS: 118-55-8 | C13H10O3 | 214.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20686.36500 g
Catalog number ALFB20686.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenyl salicylate
CAS118-55-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.
Solubility
Soluble in alcohol, ether, chloroform, turpentine, acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidants, strong acids and strong bases. Store in a cool place.
RUO – Research Use Only
General References:
  1. Shinzawa, H.; Matsumoto, Y.; Tsuzuki, S.; Shimoi, Y.; Suda, H. Photo-Fries rearrangement of phenyl salicylate studied by two-dimensional infrared spectroscopy. Vib. Spectrosc. 2015, 81, 131-135.
  2. Sen, P. K.; Chatterjee, P.; Pal, B. Evidence of co-operativity in the pre-micellar region in the hydrolytic cleavage of phenyl salicylate in the presence of cationic surfactants of CTAB, TTAB and CPC. J. Mol. Catal. A: Chem. 2015, 396, 23-30.