3-Amino-4-methoxybenzoic acid, 98+%
3-Amino-4-methoxybenzoic acid, 98+%
3-Amino-4-methoxybenzoic acid, 98+%
3-Amino-4-methoxybenzoic acid, 98+%
Thermo Scientific Chemicals
3-Amino-4-methoxybenzoic acid, 98+%
CAS: 2840-26-8 | C8H9NO3 | 167.164 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20669.065 g
Catalog number ALFB20669.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Amino-4-methoxybenzoic acid
CAS2840-26-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Yasuto Tsuruta.; Kazuya Kohashi. Sensitive derivatization reagents for hydroxyl and amino compounds for thin-layer or high-performance liquid chromatography with fluorescence detection. Anal. Chim. Acta. 1987, 192 309-313 .
  2. Manuel J. S. Monte.; Ana R. R. P. Almeida.; M. Agostinha R. Matos. Thermodynamic Study on the Sublimation of Five Aminomethoxybenzoic Acids.J. Chem. Eng. Data. 2010, 55 (1),419-423 .