(-)-Carveol, mixture of isomers, 97%
(-)-Carveol, mixture of isomers, 97%
(-)-Carveol, mixture of isomers, 97%
(-)-Carveol, mixture of isomers, 97%
Thermo Scientific Chemicals
(-)-Carveol, mixture of isomers, 97%
CAS: 99-48-9 | C10H16O | 152.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20636.0910 g
Catalog number ALFB20636.09
Price (MYR)
445.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
445.00
EA
Add to cart
Specifications
Chemical Name or Material(-)-Carveol
Name Notemixture of isomers
CAS99-48-9
Health Hazard 1H315-H320-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is a terpene that inhibits mammary carcinogenesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a terpene that inhibits mammary carcinogenesis.
Solubility
Partly soluble in water. Soluble in alcohol, ethanol and DMSO.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Carla C.C.R de Carvalho.; M.Manuela R da Fonseca. Maintenance of cell viability in the biotransformation of (-)-carveol with whole cells of Rhodococcus erythropolis. J. Mol. Catal. B: Enzym. 2002, 19-20 389-398 .
  2. Fabíola Maria N. Nunes.; M.C.C. Velosoa.; P.A. de P. Pereira.; J.B. de Andrade. Gas-phase ozonolysis of the monoterpenoids (S)-(+)-carvone, (R)-(-)-carvone, (-)-carveol, geraniol and citral. Atmos. Environ. 2005, 39 (40),7715-7730 .