Periodic acid, 99%
Periodic acid, 99%
Periodic acid, 99%
Thermo Scientific Chemicals
Periodic acid, 99%
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFB20433.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPeriodic acid
pH1.2
CAS10450-60-9
Melting Point126°C to 130°C
Health Hazard 1H271-H314-H335-H372
View more
Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.
Solubility
Soluble in water and alcohols.
Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, strong bases and dimethyl sulfoxide.
RUO – Research Use Only
General References:
  1. Oxidizing agent. For reactions of periodate ion, see Sodium metaperiodate, 13798. Reagent for deblocking of dithioacetal and oxathioacetal protecting groups under mild, non-aqueous conditions in THF-ether: Tetrahedron Lett., 37, 4331 (1996). Stoichiometric oxidant used in combination with catalytic Chromium(VI) oxide, 12522, for conversion of alcohols to carboxylic acids in wet acetonitrile: Tetrahedron Lett., 39, 5323 (1998).
  2. Yang, Z.; Sun, B.; Zhao, X.; Shao, B.; An, J.; Gu, Q.; Wang, Y.; Dong, X.; Zhang, Y.; Qiu, Z. Erythropoietin and erythropoietin receptor in hepatocellular carcinoma: correlation with vasculogenic mimicry and poor prognosis. Int J Clin Exp Pathol 2015, 8 (4), 4033-4043.
  3. Jarrett, B. R.; Gustafsson, B.; Kukis, D. L.; Louie, A. Y. Synthesis of 64Cu-labeled magnetic nanoparticles for multimodal imaging. Bioconjugate Chem. 2008, 19 (7), 1496-1504.