1-Cyclohexylethanol, 98%
1-Cyclohexylethanol, 98%
1-Cyclohexylethanol, 98%
1-Cyclohexylethanol, 98%
Thermo Scientific Chemicals
1-Cyclohexylethanol, 98%
CAS: 1193-81-3 | C8H16O | 128.215 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20363.065 g
Catalog number ALFB20363.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Cyclohexylethanol
CAS1193-81-3
Health Hazard 1H227-H312-H315-H319
Health Hazard 2GHS H Statement
H301-H311-H227
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Health Hazard 3P210-P264b-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c