4-Methyl-3-heptanol, erythro + threo, 99%
4-Methyl-3-heptanol, erythro + threo, 99%
4-Methyl-3-heptanol, erythro + threo, 99%
4-Methyl-3-heptanol, erythro + threo, 99%
4-Methyl-3-heptanol, erythro + threo, 99%
CAS: 14979-39-6 | C8H18O | 130.231 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20357.031 g
Catalog number ALFB20357.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Methyl-3-heptanol
Name Noteerythro + threo
CAS14979-39-6
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in ethanol, ether, acetone. Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. S. Steghaus-Kovâc; U. Maschwitz; A. B. Attygalle; R. T. S. Frighetto; N. Frighetto; O. Vostrowsky; H. J. Bestmann. Trail-following responses ofLeptogenys diminuta to stereoisomers of 4-methyl-3-heptanol. Experientia. 1992, 42 (7), 690-694.
  2. R.G. Riley1; R.M. Silverstein; J.C. Moser. Isolation, identification, synthesis and biological activity of volatile compounds from the heads of Atta ants. Journal of Insect Physiology. 1974, 20 (8), 1629-1637.