1,2-Dibromobutane, 98%
1,2-Dibromobutane, 98%
1,2-Dibromobutane, 98%
1,2-Dibromobutane, 98%
Thermo Scientific Chemicals
1,2-Dibromobutane, 98%
CAS: 533-98-2 | C4H8Br2 | 215.92 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20356.30250 g
Catalog number ALFB20356.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1,2-Dibromobutane
CAS533-98-2
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362