Orcein, for analysis
Orcein, for analysis
Orcein, for analysis
Orcein, for analysis
Thermo Scientific Chemicals
Orcein, for analysis
Red dye used in microscopy as a mordant for the staining of flagella, as well as for detecting elastic tissue in sputum. | CAS: 1400-62-0 | C28H24N2O7 | 500.507 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20294.031 g
Catalog number ALFB20294.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialOrcein
CAS1400-62-0
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c
Recommended StorageAmbient temperatures
View more
It is a red dye used in microscopy as a mordant for the staining of flagella, as well as for detecting elastic tissue in sputum.