cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals
cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals
cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals
cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals
cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB20288.1425 g
Catalog number ALFB20288.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialcis-3-Methylcyclohexanol
CAS24965-90-0
Recommended StorageAmbient temperatures
TSCANo
Flash Point73°C (163°F)
View more
cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.
Solubility
Soluble in ethanol, ether. Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only