1-Hexene, 98%
1-Hexene, 98%
1-Hexene, 98%
Thermo Scientific Chemicals
1-Hexene, 98%
CAS: 592-41-6 | C6H12 | 84.162 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20271.AE100 mL
Catalog number ALFB20271.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Hexene
CAS592-41-6
Health Hazard 1H225-H304-H336
Health Hazard 2GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
View more
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Hexene is used as a solvent, paint thinner and a medium for conducting chemical reactions. It is a commoner and used in the preparation of both low density and high density polyethylenes. It acts as an adhesives, lubricants, lubricant additives and sealant chemicals. Further, it is used in the preparation of hex-1-en-3-one, heptanal and heptanoic acid. It is also employed in the synthesis of linear plasticizers, oxo-alcohols, motor fuels, automotive additives and biodegradable surfactants. In addition to this, it is used in the preparation of mercaptans, flavors and fragrances, alkyl metals, halides and alkyl silanes.
Solubility
Miscible with water, ethanol, ethyl ether, benzene and petroleum ether.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Nawdiyal, A.; Hansen, N.; Zeuch, T.; Seidel, L.; Mauss, F. Experimental and modelling study of speciation and benzene formation pathways in premixed 1-hexene flames. Proc. Combust. Inst. 2015, 35 (1), 325-332.
  2. Despagnet-Ayoub, E.; Takase, M. K.; Henling, L. M.; Labinger, J. A.; Bercaw, J. E. Mechanistic Insights on the Controlled Switch from Oligomerization to Polymerization of 1-Hexene Catalyzed by an NHC-Zirconium Complex. Organometallics 2015, 34 (19), 4707-4716.