Benzoguanamine, 99%
Benzoguanamine, 99%
Benzoguanamine, 99%
Thermo Scientific Chemicals
Benzoguanamine, 99%
CAS: 91-76-9 | C9H9N5 | 187.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20157.22100 g
Catalog number ALFB20157.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzoguanamine
CAS91-76-9
Health Hazard 1H302+H332
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c