Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Tetramethylthiuram disulfide, 97%
Thermo Scientific Chemicals
Tetramethylthiuram disulfide, 97%
CAS: 137-26-8 | C6H12N2S4 | 240.416 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20063.22100 g
Catalog number ALFB20063.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylthiuram disulfide
CAS137-26-8
Health Hazard 1H302-H315-H317-H319-H373-H500
Health Hazard 2GHS H Statement
H373-H302-H332-H315-H319-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P314-P330-P333+P313-P362-P501c