4-(Trifluoromethoxy)phenyl isocyanate, 97%
4-(Trifluoromethoxy)phenyl isocyanate, 97%
4-(Trifluoromethoxy)phenyl isocyanate, 97%
4-(Trifluoromethoxy)phenyl isocyanate, 97%
Thermo Scientific Chemicals
4-(Trifluoromethoxy)phenyl isocyanate, 97%
CAS: 35037-73-1 | C8H4F3NO2 | 203.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20033.065 g
Catalog number ALFB20033.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Trifluoromethoxy)phenyl isocyanate
CAS35037-73-1
Health Hazard 1H227-H302-H311-H314-H317-H330-H334-H335
Health Hazard 2GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
View more
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethoxy)phenyl isocyanate is used as a synthetic intermediate for medicine
Solubility
Reacts with water.
Notes
Moisture Sensitive. Protect from heat. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas. Incompatible with alcohols, amines and heat.
RUO – Research Use Only