Farnesyl acetate, mixture of isomers, 96%
Farnesyl acetate, mixture of isomers, 96%
Farnesyl acetate, mixture of isomers, 96%
Farnesyl acetate, mixture of isomers, 96%
Thermo Scientific Chemicals
Farnesyl acetate, mixture of isomers, 96%
CAS: 29548-30-9 | C17H28O2 | 264.409 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19778.22100 g
Catalog number ALFA19778.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFarnesyl acetate
Name Notemixture of isomers
CAS29548-30-9
Recommended StorageAmbient temperatures
Density0.91