2,2,4-Trimethylpentane, 99%
2,2,4-Trimethylpentane, 99%
2,2,4-Trimethylpentane, 99%
2,2,4-Trimethylpentane, 99%
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99%
CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19756.0F2500 mL
Catalog number ALFA19756.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Specifications
Chemical Name or Material2,2,4-Trimethylpentane
CAS540-84-1
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
View more
2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.
Solubility
Miscible with benzene, toluene, xylene, chloroform, ether, carbon disulfide, carbon tetrachloride, DMF, acetone, heptane, and oils. Immiscible with water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Elsayed, Y.; Dallas, A.; Beatty, P. Experimental and predicted adsorption isotherms of 2,2,4-trimethylpentane and toluene on activated carbon for industrial applications. Res. Chem. Intermed. 2015, 41 (3), 1267-1282.
  2. Martins, V. D.; Granato, M. A.; Rodrigues, A. E. Isobaric Vapor-Liquid Equilibrium for Binary Systems of 2, 2, 4-Trimethylpentane with o-Xylene, m-Xylene, p-Xylene, and Ethylbenzene at 250 kPa. J. Chem. Eng. Data. 2014, 59 (5), 1499-1506.