Arachidic acid, 98%
Arachidic acid, 98%
Arachidic acid, 98%
Thermo Scientific Chemicals
Arachidic acid, 98%
CAS: 506-30-9 | C20H40O2 | 312.54 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19616.031 g
Catalog number ALFA19616.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialArachidic acid
CAS506-30-9
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.
Solubility
Soluble in chloroform, ether, ethanol, and DMF. Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Jianping Yang.; Fiona C. Meldrum.; Janos H. Fendler. Epitaxial Growth of Size-Quantized Cadmium Sulfide Crystals Under Arachidic Acid Monolayers. J. Phys. Chem. 1995, 95, (15), 5500-5504
  2. K. Kjaer.; J. Als-Nielsen.; C. A. Helm.; P. Tippman-Krayer.; H. Moehwald. Synchrotron x-ray diffraction and reflection studies of arachidic acid monolayers at the air-water interface. J. Phys. Chem. 1989, 93, (8), 3200-3206