2-Amino-4-methylphenol, 98%
2-Amino-4-methylphenol, 98%
2-Amino-4-methylphenol, 98%
2-Amino-4-methylphenol, 98%
Thermo Scientific Chemicals
2-Amino-4-methylphenol, 98%
CAS: 95-84-1 | C7H9NO | 123.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19546.1850 g
Catalog number ALFA19546.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-4-methylphenol
CAS95-84-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c