Soda lime, indicator grade
Soda lime, indicator grade
Soda lime, indicator grade
Soda lime, indicator grade
Thermo Scientific Chemicals
Soda lime, indicator grade
CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19352.36500 g
Catalog number ALFA19352.36
Price (MYR)
389.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
389.00
EA
Add to cart
Specifications
Chemical Name or MaterialSoda lime
CAS8006-28-8
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Soda lime′s ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.
Solubility
Soluble in water.
Notes
Air sensitive and hygroscopic. Incompatible with acids, water, fluorine, chloroform and trichloroethylene. It is a noncombustible material. Generates heat on hydrolysis.
RUO – Research Use Only
General References:
  1. Miller, M. B.; Gustin, M. S. Gas-exchange chamber analysis of elemental mercury deposition/emission to alluvium, ore, and mine tailings. Chemosphere 2015, 131, 209-216.
  2. Ponsot, I.; Bernardo, E.; Bontempi, E.; Depero, L.; Detsch, R.; Chinnam, R. M.; Boccaccini, A. R. Gas-exchange chamber analysis of elemental mercury deposition/emission to alluvium, ore, and mine tailings. J Clean. Prod. 2015, 89, 224-230.