Tri-n-butyl borate, 98%
Tri-n-butyl borate, 98%
Tri-n-butyl borate, 98%
Tri-n-butyl borate, 98%
Tri-n-butyl borate, 98%
Thermo Scientific Chemicals
Tri-n-butyl borate, 98%
CAS: 688-74-4 | C12H27BO3 | 230.155 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19322.AE100 mL
Catalog number ALFA19322.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-butyl borate
CAS688-74-4
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c