Perfluorotributylamine, Mass Spec Std, Thermo Scientific Chemicals
Perfluorotributylamine, Mass Spec Std, Thermo Scientific Chemicals
Perfluorotributylamine, Mass Spec Std, Thermo Scientific Chemicals
Perfluorotributylamine, Mass Spec Std, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perfluorotributylamine, Mass Spec Std, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA19126.031 g
Catalog number ALFA19126.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPerfluorotributylamine
Name NoteMass Spec Standard
CAS311-89-7
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Perfluorotributylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as a solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluorotributylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as a solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.
Solubility
Miscible with chloroform, cyclohexane, ethyl ether, ethyl acetate, heptane, carbon tetrachloride and petroleum ether. Immiscible with water, methanol, toluene, isopropyl alcohol, benzene and acetone.
Notes
Store in a cool place. Incompatible with powdered metals and alkali metals.
RUO – Research Use Only
General References:
  1. Standard for use in mass spectrometry.
  2. Wijaya, O.; Hartmann, P.; Younesi, R.; Markovits, I. I.; Rinaldi, A.; Janek, J.; Yazami, R. A gamma fluorinated ether as an additive for enhanced oxygen activity in Li-O2 batteries. J. Mater. Chem. A 2015, 3 (37), 19061-19067.
  3. Cosco, D.; Fattal, E.; Fresta, M.; Tsapis, N. Perfluorocarbon-loaded micro and nanosystems for medical imaging: A state of the art. J. Fluorine Chem. 2015, 171, 18-26.