trans-2-Octenal, 95%
trans-2-Octenal, 95%
trans-2-Octenal, 95%
trans-2-Octenal, 95%
trans-2-Octenal, 95%
CAS: 2548-87-0 | C8H14O | 126.199 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18830.065 g
Catalog number ALFA18830.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-2-Octenal
CAS2548-87-0
Health Hazard 1H227-H315-H317-H319
Health Hazard 2GHS H Statement
H315-H317-H227
Causes skin irritation.
May cause an allergic skin reaction.
Combustible liquid.
Health Hazard 3P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
View more
Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in flavoring tangerine, mandarin, cherry, melon and orange and also as a fragrances agent. It causes defects for locomotion in green fluorescent protein (GFP). It is an antimicrobial agent that destroys fungi by suppressing their ability to grow or reproduce.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and fixed oils.
Notes
Air Sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Valdemir M. Carvalho, et. al. 1,N 6-Etheno-2’-Deoxyadenosine Adducts from Trans, Trans-2,4-Decadienal and Trans-2-Octenal.Adv Exp Med Biol.2001,5000229-32.
  2. K Hasegawa.; K Fujimoto.; T Kaneda.; E N Frankel. Characterization of fluorescent products from reaction of methyl linoleate hydroperoxides with adenine in the presence of Fe2+ and ascorbic acid.Biochim Biophys Acta.1988,96(3), 371-376.