(+/-)-2-Butanol, 99%
(+/-)-2-Butanol, 99%
(+/-)-2-Butanol, 99%
(+/-)-2-Butanol, 99%
Thermo Scientific Chemicals
(+/-)-2-Butanol, 99%
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFA18658.AP
Price (MYR)
288.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
288.00
EA
Add to cart
Specifications
Chemical Name or Material(+/-)-2-Butanol
Melting Point-115°C
CAS78-92-2
Health Hazard 1H226-H319-H335-H336
Health Hazard 2GHS H Statement
H226-H319-H335-H336
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
View more
(±)-2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(^+)-2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.
Solubility
Miscible with methanol and diethyl ether. Slightly miscible with water.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Liu, H.; Li, S.; Zheng, Z.; Xu, J.; Yao, M. Effects of n-butanol, 2-butanol, and methyl octynoate addition to diesel fuel on combustion and emissions over a wide range of exhaust gas recirculation (EGR) rates. Appl. Energy 2013, 112 246-256.
  2. Togbe, C.; Mze-Ahmed, A.; Dagaut, P. Kinetics of Oxidation of 2-Butanol and Isobutanol in a Jet-Stirred Reactor: Experimental Study and Modeling Investigation. Energy Fuels 2010, 24 (9), 5244-5256.
  3. Zaitri, L. K.; Negadi, L.; Mokbel, I.; Msakni, N.; Jose, J. Liquid-vapor equilibria of binary systems containing alcohols (1-butanol, or 2-butanol or 1-hexanol) present in the production by chemical process of 2,5-dimethyl furan from biomass. Fuel 2012, 95, 438-445.