Uridine-5'-monophosphate disodium salt, 99%
Uridine-5'-monophosphate disodium salt, 99%
Uridine-5'-monophosphate disodium salt, 99%
Uridine-5'-monophosphate disodium salt, 99%
Thermo Scientific Chemicals
Uridine-5'-monophosphate disodium salt, 99%
CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA18601.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialUridine-5'-monophosphate disodium salt
CAS3387-36-8
Melting Point∼209°C (decomposition)
Recommended StorageKeep cold
RTECS NumberYU7975000