5-Nitro-2-furaldehyde semicarbazone, 98+%
5-Nitro-2-furaldehyde semicarbazone, 98+%
5-Nitro-2-furaldehyde semicarbazone, 98+%
5-Nitro-2-furaldehyde semicarbazone, 98+%
Thermo Scientific Chemicals
5-Nitro-2-furaldehyde semicarbazone, 98+%
CAS: 59-87-0 | C6H6N4O4 | 198.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18593.1425 g
Catalog number ALFA18593.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Nitro-2-furaldehyde semicarbazone
CAS59-87-0
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H301-H341-H351-H361
Toxic if swallowed.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
View more
Bactericidal compound used as an antibiotic