D-Tryptophan, 99%
D-Tryptophan, 99%
D-Tryptophan, 99%
Thermo Scientific Chemicals
D-Tryptophan, 99%
CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA18426.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Tryptophan
Melting Point273°C (decomposition)
CAS153-94-6
Recommended StorageAmbient temperatures
RTECS NumberYN6129000
View more
D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Tryptophan is used for human embryonic kidney cell (HEK-293, ATCC: CRL-1573) culture. The product is a sweetener used to study the release of incretins from enteroendocrine cells triggered by sugar but not by sweeteners.
Solubility
Soluble in 0.5M hydrochloric acid (50 mg/ml), water (11 mg/ml at 20°C), hot alcohol, and dimethyl sulfoxide. Insoluble in chloroform.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Yukihiro Fujita.; Rhonda D Wideman.; Madeleine Speck.; Ali Asadi.; David S King.; Travis D Webber.; Masakazu Haneda.; Timothy J Kieffer. Incretin release from gut is acutely enhanced by sugar but not by sweeteners in vivo. American Journal of Physiology - Endocrinology and Metabolism. 2009, 296 (3), 473-479.
  2. Daniela Vullo.; Alessio Innocenti.; Isao Nishimori.; Andrea Scozzafava.; Kai Kaila.; Claudiu T Supuran. Carbonic anhydrase activators: activation of the human isoforms VII (cytosolic) and XIV (transmembrane) with amino acids and amines. Bioorganic & Medicinal Chemistry Letters. 2007, 17 (15), 4107-4112.