Rhodanine, 98+%
Rhodanine, 98+%
Rhodanine, 98+%
Thermo Scientific Chemicals
Rhodanine, 98+%
CAS: 141-84-4 | C3H3NOS2 | 133.183 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18293.1425 g
Catalog number ALFA18293.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodanine
CAS141-84-4
Health Hazard 1H302-H319
Health Hazard 2GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
View more
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.
Solubility
Soluble in water (2.25 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. H.J.Eggers; M.A.Koch; A.Furst; G.D.Daves; J.J.Wilczynski; K.Folkers. Rhodanine: a selective inhibitor of the multiplication of echovirus 12. Science. 1970, 167,(3916), 294-297.
  2. W.T.Sing; C.L.Lee; S.L.Yeo; S.P.Lim; M.M.Sim. Arylalkylidene rhodanine with bulky and hydrophobic functional group as selective HCV NS3 protease inhibitor. Bioorganic & Medicinal Chemistry Letters. 2001, 11,(2), 91-94.
  3. Reagent for Ag and Au.