3-Aminobenzeneboronic acid monohydrate, 97%
3-Aminobenzeneboronic acid monohydrate, 97%
3-Aminobenzeneboronic acid monohydrate, 97%
3-Aminobenzeneboronic acid monohydrate, 97%
Thermo Scientific Chemicals
3-Aminobenzeneboronic acid monohydrate, 97%
CAS: 206658-89-1 | C6H10BNO3 | 154.96 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18189.065 g
Catalog number ALFA18189.06
Price (MYR)
624.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
624.00
EA
Add to cart
Specifications
Chemical Name or Material3-Aminobenzeneboronic acid monohydrate
CAS206658-89-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is employed as a reagent in the preparation of Suzuki-Miyaura cross-coupling, used for Gram-positive ant virulence drugs and inhibitors of Streptococcus agalactiae Stk1, regioisomer of Zaleplon (a sedative), amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix, chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity. Aminophenylboronic Acid is also used as a reagent in the preparation of nonbenzodiazepine hypnotic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a reagent in the preparation of Suzuki-Miyaura cross-coupling, used for Gram-positive ant virulence drugs and inhibitors of Streptococcus agalactiae Stk1, regioisomer of Zaleplon (a sedative), amphiphilic random glycopolymer, which self-assemble to form nanoparticles, with potential as a glucose-sensitive matrix, chemomechanical polymer that expands and contracts in blood plasma with high glucose selectivity. Aminophenylboronic Acid is also used as a reagent in the preparation of nonbenzodiazepine hypnotic agents.
Solubility
Soluble in DMSO and methanol.
Notes
Air & Moisture Sensitive. Store away from oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Gregory D. Cuny.; Maxime Robin.; Natalia P. Ulyanova.; Debasis Patnaik.; Valerie Pique.; Gilles Casano.; Ji-Feng Liu.; Xiangjie Lin.; Jun Xian.; Marcie A. Glicksman.; Ross L. Stein.; Jonathan M.G. Higgins. Structure-activity relationship study of acridine analogs as haspin and DYRK2 kinase inhibitors.Bioorg Med Chem Lett.2010,20(12), 3491-3494.
  2. Xingju Jin.; Xinge Zhang.; Zhongming Wu.; Dayong Teng.; Xuejiao Zhang.; Yanxia Wang.; Zhen Wang.; Chaoxing Li. Amphiphilic random glycopolymer based on phenylboronic acid: synthesis, characterization, and potential as glucose-sensitive matrix.Biomacromolecules.2009,10(6), 1337-1345.