Hydrocortisone acetate, 97+%
Hydrocortisone acetate, 97+%
Hydrocortisone acetate, 97+%
Hydrocortisone acetate, 97+%
Thermo Scientific Chemicals
Hydrocortisone acetate, 97+%
CAS: 50-03-3 | C23H32O6 | 404.50 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA18089.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrocortisone acetate
CAS50-03-3
Health Hazard 1H360-H373
Health Hazard 2GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3P201-P202-P260-P281-P308+P313-P501c
View more
Hydrocortisone acetate is used to treat hemorrhoids and itching/swelling in the rectum and anus. It is used to treat certain intestinal problems (such as ulcerative colitis of the rectum and other rectal/anal inflammatory conditions).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store in the dark.
RUO – Research Use Only
Dan-Ni Wang; Takahito Toyotome; Yasunori Muraosa; Akira Watanabe; Tuya Wuren; Somnuk Bunsupa; Kaori Aoyagi; Mami Yamazaki; Masahiko Takino; Katsuhiko Kamei. GliA in Aspergillus fumigatus is required for its tolerance to gliotoxin and affects the amount of extracellular and intracellular gliotoxin. Medical Mycology.2014, 52, (5), 506-518.