(+/-)-Camphene, tech. (sum of camphene + fenchene)
(+/-)-Camphene, tech. (sum of camphene + fenchene)
(+/-)-Camphene, tech. (sum of camphene + fenchene)
(+/-)-Camphene, tech. (sum of camphene + fenchene)
Thermo Scientific Chemicals
(+/-)-Camphene, tech. (sum of camphene + fenchene)
CAS: 79-92-5 | C10H16 | 136.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18040.1850 g
Catalog number ALFA18040.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-Camphene
Name Notesum of camphene + fenchene
CAS79-92-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H228-H319
Flammable solid.
Causes serious eye irritation.
View more
Camphene is used in the preparation of fragrances and as a food additive for flavoring.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Camphene is used in the preparation of fragrances and as a food additive for flavoring.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Eun-Jung Lee, et al. Highly porous hydroxyapatite bioceramics with interconnected pore channels using camphene-based freeze casting.Materials Lett.2007,61(11-12), 2270-2273.
  2. B Bayender, et al. Solid-liquid interfacial energy of camphene.Mater. Sci. Eng., A.1999,270(2), 343-348.