Di-n-octylamine, 96%
Di-n-octylamine, 96%
Di-n-octylamine, 96%
Di-n-octylamine, 96%
Thermo Scientific Chemicals
Di-n-octylamine, 96%
CAS: 1120-48-5 | C16H35N | 241.463 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18017.22100 g
Catalog number ALFA18017.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-octylamine
CAS1120-48-5
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.
Solubility
Immiscible in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only
General References:
  1. R Stocker; Y Yamamoto; AF McDonagh; AN Glazer; BN Ames. Bilirubin is an antioxidant of possible physiological importance. Science. 1987, 235 (4792), 1043-1046.
  2. S. L. Oswal; A. T. Patel. Viscosity of binary mixtures. I. mono-, di-, and tri-n-butyl and -n-octylamine with cyclohexane. International Journal of Thermophysics. 1991, 12 (5), 821-835.